Learn More
Abcam PD 151746, Calpain inhibitor, 25MG
Catalog No. 502705145
Encompass_Preferred
MW 237.25 g/mol, Purity >96%. Cell permeable, selective, non-peptidic calpain inhibitor (Ki values are 0.3 and 5 μM for calpain 1 and calpain 2, respectively). Stimulates phagocyte function via hFPR/hFPRL1 activation. Interacts with the Ca²+ calpain sites. Shows antiapoptotic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5145 Supplier Abcam Supplier No. AB14552325MG
MW 237.25 g/mol, Purity >96%. Cell permeable, selective, non-peptidic calpain inhibitor (Ki values are 0.3 and 5 μM for calpain 1 and calpain 2, respectively). Stimulates phagocyte function via hFPR/hFPRL1 activation. Interacts with the Ca²+ calpain sites. Shows antiapoptotic effects.
The product is subject to the following: Abcam Restricted Use Statement