Learn More
Cayman Chemical PD 153035 (hydrochloride), Purity: ≥98%, CAS: 183322-45-4, 50 mg
SDP
Catalog No. 502969730
Encompass_Preferred
A highly potent, reversible inhibitor of the EGFR (Ki = 5.2 pM; IC50 = 29 pM); rapidly suppresses autophosphorylation of EGFR in fibroblasts and human epidermoid carcinoma cells
Catalog No. 50-296-9730 Supplier Cayman Chemical Supplier No. 1487950 MG
A highly potent, reversible inhibitor of the EGFR (Ki = 5.2 pM; IC50 = 29 pM); rapidly suppresses autophosphorylation of EGFR in fibroblasts and human epidermoid carcinoma cells