Learn More
Medchemexpress LLC HY-107428 5mg Medchemexpress, PD-166793 CAS:199850-67-4 Purity:>98%
SDP
Catalog No. 502080711
Encompass_Preferred
Medchemexpress, HY-107428 5mg PD-166793 CAS:199850-67-4 PD-166793 is a potent, selective, orally active and wide‐broad spectrum inhibitor of MMP, exhibiting nanomolar potency against MMP-2, MMP-3 and MMP-13 (IC50=4, 7, and 8 nM, respectively) and micromolar potency vs MMP-1, -7 and -9 (IC50=6.0, 7.2, and 7.9 μM, respectively). PD-166793 can attenuate left ventricular remodeling and dysfunction in a rat model of progressive heart failure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0711 Supplier Medchemexpress LLC Supplier No. HY1074285MG
Medchemexpress, HY-107428 5mg PD-166793 CAS:199850-67-4 PD-166793 is a potent, selective, orally active and wide‐broad spectrum inhibitor of MMP, exhibiting nanomolar potency against MMP-2, MMP-3 and MMP-13 (IC50=4, 7, and 8 nM, respectively) and micromolar potency vs MMP-1, -7 and -9 (IC50=6.0, 7.2, and 7.9 μM, respectively). PD-166793 can attenuate left ventricular remodeling and dysfunction in a rat model of progressive heart failure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.