Learn More
Medchemexpress LLC HY-101296 5mg Medchemexpress, PD-166866 CAS:192705-79-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012965MG
Encompass_Preferred
Medchemexpress, HY-101296 5mg PD-166866 CAS:192705-79-6 PD166866 is a selective FGFR1 tyrosine kinase inhibitor with an IC 50 of 52.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2090