Learn More
Medchemexpress LLC Pd-168077 maleate | 630117-19-0 | 99.6% | 450.49 | 1 ML
SDP
Catalog No. 5000400924
Encompass_Preferred
PD-168077 maleate is a selective dopamine D4 receptor agonist with a Ki of 9 nM. It is intended for research use only. It exhibits intrinsic activity at the D4 receptor, leading to quinpirole-like inhibition of forskolin-stimulated cAMP accumulation or stimulation of [3H]thymidine uptake.
  • Shows >100-fold selectivity over other D2-like receptor family members and D1-like counterparts.
  • Demonstrates 20-fold selectivity over α1 and α2 receptors.
  • Exhibits 45-fold selectivity over 5-HT1A.
  • Shows 460-fold selectivity over 5-HT2A receptors.
  • Induces synaptic translocation of Ca2+/calmodulin-dependent protein kinase II in cultured prefrontal cortical neurons.
Catalog No. 50-004-00924 Supplier Medchemexpress LLC Supplier No. HY21098A10MM/1ML
PD-168077 maleate is a selective dopamine D4 receptor agonist with a Ki of 9 nM. It is intended for research use only. It exhibits intrinsic activity at the D4 receptor, leading to quinpirole-like inhibition of forskolin-stimulated cAMP accumulation or stimulation of [3H]thymidine uptake.
  • Shows >100-fold selectivity over other D2-like receptor family members and D1-like counterparts.
  • Demonstrates 20-fold selectivity over α1 and α2 receptors.
  • Exhibits 45-fold selectivity over 5-HT1A.
  • Shows 460-fold selectivity over 5-HT2A receptors.
  • Induces synaptic translocation of Ca2+/calmodulin-dependent protein kinase II in cultured prefrontal cortical neurons.