Learn More
Medchemexpress LLC PD-168077 maleate | 630117-19-0 | 99.6% | 450.49 | 100 MG
SDP
Catalog No. 5000401182
Encompass_Preferred
PD-168077 maleate is a selective dopamine D4 receptor agonist with a Ki of 9 nM. It is for research use only. It shows high selectivity over other D2-like receptor family members and D1-like counterparts, as well as α1, α2, 5-HT1A, and 5-HT2A receptors. It evidences intrinsic activity at the D4 receptor and can induce locomotion in a dose-dependent manner. This product may be used in research related to neurological diseases, neurodegenerative diseases, depression, and Parkinson's disease.
  • Selective dopamine D4 receptor agonist
  • High selectivity over other receptor types
  • Evidences intrinsic activity at the D4 receptor
  • Induces locomotion in PD-168077-treated cells
  • May be used in research for neurological diseases
Catalog No. 50-004-01182 Supplier Medchemexpress LLC Supplier No. HY21098A100MG
PD-168077 maleate is a selective dopamine D4 receptor agonist with a Ki of 9 nM. It is for research use only. It shows high selectivity over other D2-like receptor family members and D1-like counterparts, as well as α1, α2, 5-HT1A, and 5-HT2A receptors. It evidences intrinsic activity at the D4 receptor and can induce locomotion in a dose-dependent manner. This product may be used in research related to neurological diseases, neurodegenerative diseases, depression, and Parkinson's disease.
  • Selective dopamine D4 receptor agonist
  • High selectivity over other receptor types
  • Evidences intrinsic activity at the D4 receptor
  • Induces locomotion in PD-168077-treated cells
  • May be used in research for neurological diseases