Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PD-168077 maleate | 630117-19-0 | 99.6% | 450.49 | 50 MG
SDP

Catalog No. 5000401181
Encompass_Preferred

PD-168077 maleate is a selective dopamine D4 receptor agonist with a Ki of 9 nM. It exhibits high selectivity over other D2-like and D1-like receptor family members, as well as α1, α2, 5-HT1A, and 5-HT2A receptors. This compound shows intrinsic activity at the D4 receptor, inhibiting forskolin-stimulated cAMP accumulation or stimulating [3H]thymidine uptake in CHO cells expressing the human D4 receptor. In vivo, PD-168077 dose-dependently induces locomotion.

  • Selective dopamine D4 receptor agonist
  • Exhibits high selectivity over other D2-like, D1-like, α1, α2, 5-HT1A, and 5-HT2A receptors
  • Shows intrinsic activity at the D4 receptor
  • Induces locomotion in vivo

Catalog No. 50-004-01181 Supplier Medchemexpress LLC Supplier No. HY21098A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PD-168077 maleate is a selective dopamine D4 receptor agonist with a Ki of 9 nM. It exhibits high selectivity over other D2-like and D1-like receptor family members, as well as α1, α2, 5-HT1A, and 5-HT2A receptors. This compound shows intrinsic activity at the D4 receptor, inhibiting forskolin-stimulated cAMP accumulation or stimulating [3H]thymidine uptake in CHO cells expressing the human D4 receptor. In vivo, PD-168077 dose-dependently induces locomotion.

  • Selective dopamine D4 receptor agonist
  • Exhibits high selectivity over other D2-like, D1-like, α1, α2, 5-HT1A, and 5-HT2A receptors
  • Shows intrinsic activity at the D4 receptor
  • Induces locomotion in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.