Learn More
Abcam PD 168077 maleate, D4 agonist, 10MG
Catalog No. 502707474
Encompass_Preferred
MW 450.49 g/mol, Purity >98%. Potent, selective D₄ dopamine receptor agonist. Displays >400-fold selectivity over D₂ and >300-fold selectivity over D₃ subtypes (Ki values are 11.9, 2540, and 1050 nM for D₄, D₃, and D₂ receptors, respectively). Reverses object recognition deficits.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7474 Supplier Abcam Supplier No. AB12056810MG
MW 450.49 g/mol, Purity >98%. Potent, selective D₄ dopamine receptor agonist. Displays >400-fold selectivity over D₂ and >300-fold selectivity over D₃ subtypes (Ki values are 11.9, 2540, and 1050 nM for D₄, D₃, and D₂ receptors, respectively). Reverses object recognition deficits.
The product is subject to the following: Abcam Restricted Use Statement