Learn More
Selleck Chemical LLC PD 169316-5mg
SDP
Catalog No. 502389911
Encompass_Preferred
PD 169316 is a potent, selective and cell-permeable p38 MAP kinase inhibitor with IC50 of 89 nM. PD169316 abrogates signaling initiated by both TGFbeta and Activin A. PD169316 shows antiviral activity against Enterovirus71.
Catalog No. 50-238-9911 Supplier Selleck Chemical LLC Supplier No. S51835MG
PD 169316 is a potent, selective and cell-permeable p38 MAP kinase inhibitor with IC50 of 89 nM. PD169316 abrogates signaling initiated by both TGFbeta and Activin A. PD169316 shows antiviral activity against Enterovirus71.