Learn More
Apexbio Technology LLC PD 173074, 50mg. Cas: 219580-11-7 MFCD: MFCD08705327. FGFR inhibitor
SDP
Supplier:  Apexbio Technology LLC A825350
Encompass_Preferred
PD173074 is a poten and selective fibroblast growth factor receptor (FGFR) inhibitor. Fibroblast growth factor-2 is reported to be able to induce proliferation and chemoresistance in Small cell lung cancer (SCLC) cells. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5092