Learn More
Abcam PD 174265, EGFR tyrosine kinase inhibitor, 1MG
Catalog No. 502705426
Encompass_Preferred
MW 371.24 g/mol, Purity >98%. Potent, selective, reversible EGFR tyrosine kinase inhibitor (IC₅₀ = 0.45 nM). Antitumor agent. Cell permeable. Active in vivo and in vitro. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5426 Supplier Abcam Supplier No. AB1451461MG
Add to Cart
Add to Cart
MW 371.24 g/mol, Purity >98%. Potent, selective, reversible EGFR tyrosine kinase inhibitor (IC₅₀ = 0.45 nM). Antitumor agent. Cell permeable. Active in vivo and in vitro. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement