Learn More
Enzo Life Sciences PD 184,352 (1mg). CAS: 212631-79-3
SDP
Supplier:  Enzo Life Sciences ALX270471M001
Encompass_Preferred
Potent and selective inhibitor of MAPK (ERK kinase 1; MEK1) activation (IC50=300nM in vitro, IC50=2nM in vivo). Suppresses activation of MAPK but does not block its activity. Strongly suppresses growth of human implanted colon tumors in mice. Alternative name: 2-(2-Chloro-4-iodo-phenylamino)-N-cyclopropylmethoxy-3,4-difluorobenzamide, CI-1040. Purity: ≥95% (HPLC). Solubility: Soluble in organic solvents. Long Term Storage: -20°C.
Catalog No. 50-200-8597