Learn More
AdipoGen PD 184 352
SDP
Catalog No. 502562387
Encompass_Preferred
Chemical. CAS 212631-79-3. Formula C17H14ClF2IN2O2. MW 478.7. MEK MAPKK inhibitor. Potent and selective MAPK ERK kinase 1 MEK1 activation inhibitor IC50 = 300 nM in vitro, IC50 = 2 nM in vivo. Suppresses activation of MAPK but does not block its activity. Antiproliferative. Causes cell-cycle arrest in G1 phase. Tumor suppressor. Apoptosis inducer.
Catalog No. 50-256-2387 Supplier AdipoGen Supplier No. AGCR10029M001
Chemical. CAS 212631-79-3. Formula C17H14ClF2IN2O2. MW 478.7. MEK MAPKK inhibitor. Potent and selective MAPK ERK kinase 1 MEK1 activation inhibitor IC50 = 300 nM in vitro, IC50 = 2 nM in vivo. Suppresses activation of MAPK but does not block its activity. Antiproliferative. Causes cell-cycle arrest in G1 phase. Tumor suppressor. Apoptosis inducer.