Learn More
Cayman Chemical PD 184161 Inhibitors
SDP
Supplier:  Cayman Chemical 1001243125 MG
Encompass_Preferred
A potent MEK1/2 inhibitor (IC50 = 10-100 nM); more effective at inhibiting phosphorylation of ERK1/2 than PD 098059 and U0126; inhibits proliferation, induces apoptosis, and possesses antitumor activity in MEK-dependent cancers
Catalog No. 50-288-8653