Learn More
Cayman Chemical PD 184161 Inhibitors
SDP
Catalog No. 502889234
Encompass_Preferred
A potent MEK1/2 inhibitor (IC50 = 10-100 nM); more effective at inhibiting phosphorylation of ERK1/2 than PD 098059 and U0126; inhibits proliferation, induces apoptosis, and possesses antitumor activity in MEK-dependent cancers
Catalog No. 50-288-9234 Supplier Cayman Chemical Supplier No. 100124315 MG
Add to Cart
Add to Cart
A potent MEK1/2 inhibitor (IC50 = 10-100 nM); more effective at inhibiting phosphorylation of ERK1/2 than PD 098059 and U0126; inhibits proliferation, induces apoptosis, and possesses antitumor activity in MEK-dependent cancers