Learn More
Cayman Chemical PD-1/PD-L1 Inhibitor 1, Purity: ≥98%, CAS: 1675201-83-8, 25 mg
SDP
Catalog No. 502984597
Encompass_Preferred
A PD-1/PD-L1 interaction inhibitor; inhibits the PD-1/PD-L1 interaction in a TR-FRET assay (IC50 = 6-100 nM); reduces LPS-induced increases in PD-1 levels and apoptosis in MH-S mouse alveolar macrophages at 1 µM; surgical neointimal patching with PD-1/PD-L1-coated patches decreases neointimal thickness and inflammatory cell accumulation in a rat model of patch angioplasty
Catalog No. 50-298-4597 Supplier Cayman Chemical Supplier No. 1991425 MG
A PD-1/PD-L1 interaction inhibitor; inhibits the PD-1/PD-L1 interaction in a TR-FRET assay (IC50 = 6-100 nM); reduces LPS-induced increases in PD-1 levels and apoptosis in MH-S mouse alveolar macrophages at 1 µM; surgical neointimal patching with PD-1/PD-L1-coated patches decreases neointimal thickness and inflammatory cell accumulation in a rat model of patch angioplasty