Learn More
Cayman Chemical PD-1/PD-L1 Inhibitor 2, Purity: ≥98%, CAS: 1675203-84-5, 1 mg
SDP
Catalog No. 502980727
Encompass_Preferred
Prevents the interaction of PD-L1 with PD-1 with an IC50 value of 18 nM
Catalog No. 50-298-0727 Supplier Cayman Chemical Supplier No. 191321 MG
Prevents the interaction of PD-L1 with PD-1 with an IC50 value of 18 nM