Learn More
Cayman Chemical PD-1/PD-L1 Inhibitor 3 (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502909393
Encompass_Preferred
A PD-1/PD-L1 interaction inhibitor (IC50 = 9 nM in a TR-FRET assay)
Catalog No. 50-290-9393 Supplier Cayman Chemical Supplier No. 367191 MG
A PD-1/PD-L1 interaction inhibitor (IC50 = 9 nM in a TR-FRET assay)