Learn More
Cayman Chemical PD-1PD-L1 InhIbItr N11 1mg
SDP
Catalog No. 502918449
Encompass_Preferred
A PD-1/PD-L1 interaction inhibitor (IC50 = 6.3 nM); increases T cell proliferation in a PD-L1-overexpressing CT26 colon cancer cell and T cell co-culture at 20 nM; promotes T cell infiltration into an in vitro PD-L1-overexpressing murine Lewis lung carcinoma tumor matrix at 20 and 80 nM; reduces tumor growth and promotes CD8+ and CD4+ T cell tumor infiltration in a PD-L1-overexpressing murine Lewis lung carcinoma model at 20 mg/kg
Catalog No. 50-291-8449 Supplier Cayman Chemical Supplier No. 396631 MG
Add to Cart
Add to Cart
A PD-1/PD-L1 interaction inhibitor (IC50 = 6.3 nM); increases T cell proliferation in a PD-L1-overexpressing CT26 colon cancer cell and T cell co-culture at 20 nM; promotes T cell infiltration into an in vitro PD-L1-overexpressing murine Lewis lung carcinoma tumor matrix at 20 and 80 nM; reduces tumor growth and promotes CD8+ and CD4+ T cell tumor infiltration in a PD-L1-overexpressing murine Lewis lung carcinoma model at 20 mg/kg