Learn More
Medchemexpress LLC Pd 4'-oxyacetic acid | 2633633-12-0 | 99.7% | 264.24 g·mol⁻¹ | C12H12N2O5 | 100mg
SDP
Catalog No. 5000269471
Encompass_Preferred
PD 4 -oxyacetic acid is a carboxylic acid-functionalized cereblon ligand that can be used for PROTAC Degrader development[1]
Catalog No. 50-002-69471 Supplier Medchemexpress LLC Supplier No. HYW761299100MG
Add to Cart
Add to Cart
PD 4 -oxyacetic acid is a carboxylic acid-functionalized cereblon ligand that can be used for PROTAC Degrader development[1]