Learn More
Medchemexpress LLC HY-16958 5mg , PD 404182 CAS:72596-74-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY169585MG
Encompass_Preferred
HY-16958 5mg PD 404182 CAS: 72596-74-8 PD 404182 is a potent and competitive inhibitor of human dimethylarginine dimethylaminohydrolase 1 (DDAH1), with an IC50 of 9 μM. PD 404182 exhibits antiangiogenic and antiviral activity in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1893