Learn More
Nelson Jameson Company Magrabar PD-650 Non-Silicone Defoamer, 40/pk
Catalog No. NC1456064
Encompass
Use this food-grade, non-silicone defoamer, to destroy existing foam in protein and carbohydrate media. The formula is a water emulsion that strictly adheres to FDA direct/indirect ingredient approvals. Works well in a wide range of pH and temperatures in the cheese and dairy industry. Certified Kosher and Halal.
Catalog No. NC1456064 Supplier Nelson Jameson Company Supplier No. 4154040 FM
Use this food-grade, non-silicone defoamer, to destroy existing foam in protein and carbohydrate media. The formula is a water emulsion that strictly adheres to FDA direct/indirect ingredient approvals. Works well in a wide range of pH and temperatures in the cheese and dairy industry. Certified Kosher and Halal.