Learn More
TARGETMOL CHEMICALS INC PD 90780 5MG
SDP
Catalog No. 502581458
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD 90780 is a non peptide antagonist of nerve growth factor (NGF). PD 90780 interacts with NGF prevents NGF binds with p75NTR. PD 90780 inhibits NGF-p75NTR interaction with IC50s of 23.1 and 1.8 µM in PC12 cells and PC12nnr5 cells respectively. purity: 98%
Catalog No. 50-258-1458 Supplier TARGETMOL CHEMICALS INC Supplier No. T123845MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD 90780 is a non peptide antagonist of nerve growth factor (NGF). PD 90780 interacts with NGF prevents NGF binds with p75NTR. PD 90780 inhibits NGF-p75NTR interaction with IC50s of 23.1 and 1.8 µM in PC12 cells and PC12nnr5 cells respectively. purity: 98%