Learn More
Medchemexpress LLC HY-110166 5mg Medchemexpress, PD 90780 CAS:77422-99-2 Purity:>98%
SDP
Catalog No. 501932346
Encompass_Preferred
Medchemexpress, HY-110166 5mg PD 90780 CAS:77422-99-2 PD 90780 is a non peptide antagonist of nerve growth factor (NGF) binding to the P75 NGF receptor, inhibits NGF-p75 NTR interaction with IC 50 s values of 23.1 µM and 1.8 µM in PC12 cells and PC12 nnr5 cells, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2346 Supplier Medchemexpress LLC Supplier No. HY1101665MG
Medchemexpress, HY-110166 5mg PD 90780 CAS:77422-99-2 PD 90780 is a non peptide antagonist of nerve growth factor (NGF) binding to the P75 NGF receptor, inhibits NGF-p75 NTR interaction with IC 50 s values of 23.1 µM and 1.8 µM in PC12 cells and PC12 nnr5 cells, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.