Learn More
AdipoGen PD 98,059 (10 mg)
SDP
Supplier:  AdipoGen AGCR10118M010
Encompass_Preferred
Chemical. CAS: 167869-21-8. Formula: C16H13NO3. MW: 267.3. Highly selective, reversible and cell permeable MEK (MAP kinase kinase) inhibitor. Blocks the phosphorylation and activation of the MAP kinase pathway. T cell activation inhibitor. Inhibits cell growth and cell proliferation of several cancer cells.
Catalog No. 50-205-3847