Learn More
Cayman Chemical PD 98059 Inhibitors
SDP
Catalog No. 502890296
Encompass_Preferred
A MAPK pathway inhibitor and AMPK activator; prevents the activation of MAP2K1 by Raf or MEK kinase (IC50 = 2-7 µM); activator of AMPK (EC50 = 35 µM); increases the ratios of ADP:ATP and AMP:ATP
Catalog No. 50-289-0296 Supplier Cayman Chemical Supplier No. 1000672610 MG
Add to Cart
Add to Cart
A MAPK pathway inhibitor and AMPK activator; prevents the activation of MAP2K1 by Raf or MEK kinase (IC50 = 2-7 µM); activator of AMPK (EC50 = 35 µM); increases the ratios of ADP:ATP and AMP:ATP