Learn More
REPLIGEN CORPORATION Repligen PD Cassette 300KD, 0.02 m2 Membrane Area, 300 kD MWCO
SDP
Catalog No. NC3440445
Encompass
The Repligen PRO PD 0.02m2 (LP) HyS 300kD Cassette is designed for efficient tangential flow filtration in process development applications. With a HyStream membrane offering a 300 kD molecular weight cut-off and a 0.02 m2 membrane area, it provides superior ultrafiltration and diafiltration capabilities. Its unique L-type channel configuration and compatibility with holder-A systems enhance flexibility, making it suitable for both single-use and reusable bioprocess workflows.
Catalog No. NC3440445 Supplier REPLIGEN CORPORATION Supplier No. XP300AP2L
The Repligen PRO PD 0.02m2 (LP) HyS 300kD Cassette is designed for efficient tangential flow filtration in process development applications. With a HyStream membrane offering a 300 kD molecular weight cut-off and a 0.02 m2 membrane area, it provides superior ultrafiltration and diafiltration capabilities. Its unique L-type channel configuration and compatibility with holder-A systems enhance flexibility, making it suitable for both single-use and reusable bioprocess workflows.