Learn More
Strem, An Ascensus Company CAS# 225931-80-6. 250mg. Bis[(trimethylsilyl)methyl](1,5-cyclooctadiene)palladium(II), 98%.
SDP
Supplier:  Strem, An Ascensus Company 460308250MG
Encompass
CAS# 225931-80-6. 250mg. Bis[(trimethylsilyl)methyl](1,5-cyclooctadiene)palladium(II), 98%. . Molecular Weight: 389.03. Molecular Formula: C16H34PdSi2. Color/form: gray pwdr. Strem# 46-0308. http://www.strem.com/catalog/v/46-0308/
Catalog No. 50-201-6753