Learn More
Sigma Aldrich Pd Encat™ 30
Catalog No. 64471410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
64-471-410G 5 g
1 options
Danger
Catalog No. 64-471-410G Supplier Sigma Aldrich Supplier No. 64471410G
  • NEW LOWER PRICE
Danger
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
It is an encapsulated catalyst (EnCat™), wherein palladium acetate is microencapsulated within a polyurea matrix, which prevents palladiu mLeaching.While Pd EnCat 40 shows excellent performance in a variety of transformations, Pd EnCat 30, with lower matrix content, shows similar performance but with improved kinetics while maintaining the mechanically robust nature of Pd EnCat 40 under similar conditions.Matrix content 30%Pd Encat™ technology.
Specifications
CAS 129423-29-6
Quantity 5 g