Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 50mg, Pd(II) meso-Tetraphenyl Tetrabenzoporphine, >95%, [119654-64-7], 41217 919.37 C60H36N4Pd

Catalog No. 501374487
Encompass_Preferred

Compound: Pd(II) meso-Tetraphenyl Tetrabenzoporphine

CAS#: 119654-64-7

MDL#: MFCD03095471

Background: A synthetic palladium(II) porphyrin specialty chemical related to platinum tetrabenzoporphine analogs used in oxygen sensing and photophysical research. Metallated tetrabenzoporphyrins are of interest for their extended conjugation and phosphorescent properties.

Storage: Store at room temperature, protected from light.

Catalog No. 50-137-4487 Supplier Frontier Specialty Chemicals Supplier No. 4121750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Pd(II) meso-Tetraphenyl Tetrabenzoporphine

CAS#: 119654-64-7

MDL#: MFCD03095471

Background: A synthetic palladium(II) porphyrin specialty chemical related to platinum tetrabenzoporphine analogs used in oxygen sensing and photophysical research. Metallated tetrabenzoporphyrins are of interest for their extended conjugation and phosphorescent properties.

Storage: Store at room temperature, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.