Learn More
Bioss PD-L1 - Extracellular region M
SDP
Supplier:  Bioss BSM70603M
Encompass_Preferred
PD-L1 (Extracellular region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M051Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-66541