Learn More
Medchemexpress LLC HY-P7397 50ug Medchemexpress, PD-L1, Fc Chimera (CHO-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP739750UG
Encompass_Preferred
Medchemexpress, HY-P7397 50ug PD-L1, Fc Chimera (CHO-expressed), Human CAS: Recombinant Human PD-L1, Fc Chimera (CHO-expressed) play a major role in suppressing the adaptive arm of immune system during particular events such as pregnancy, tissue allografts, autoimmune disease and other disease states such as hepatitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3158