Learn More
Genscript Corporation PD-L1 Fc Chimera Mouse
Catalog No. 5000059522
Encompass
Programmed death-ligand 1 (PD-L1) also known as cluster of differentiation 274 (CD274) or B7 homolog 1 (B7-H1) is a protein that in humans is encoded by the CD274 gene PD-L1 is a 40 kDa type 1 transmembrane protein that has been speculated to play a major role in suppressing the immune system during particular events such as pregnancy tissue allografts autoimmune disease and other disease states such as hepatitis Normally the immune system reacts to foreign antigens where there is some accumulation in the lymph nodes or spleen which triggers a proliferation of antigen-specific CD8 T cell The formation of PD-1 receptor / PD-L1 or B7 1 receptor /PD-L1 ligand complex transmits an inhibitory signal which reduces the proliferation of these CD8 T cells at the lymph nodes and supplementary to that PD-1 is also able to control the accumulation of foreign antigen specific T cells in the lymph nodes through apoptosis which is further mediated by a lower regulation of the gene Bcl-2 PD-L
Catalog No. 50-000-59522 Supplier Genscript Corporation Supplier No. Z0338410
Programmed death-ligand 1 (PD-L1) also known as cluster of differentiation 274 (CD274) or B7 homolog 1 (B7-H1) is a protein that in humans is encoded by the CD274 gene PD-L1 is a 40 kDa type 1 transmembrane protein that has been speculated to play a major role in suppressing the immune system during particular events such as pregnancy tissue allografts autoimmune disease and other disease states such as hepatitis Normally the immune system reacts to foreign antigens where there is some accumulation in the lymph nodes or spleen which triggers a proliferation of antigen-specific CD8 T cell The formation of PD-1 receptor / PD-L1 or B7 1 receptor /PD-L1 ligand complex transmits an inhibitory signal which reduces the proliferation of these CD8 T cells at the lymph nodes and supplementary to that PD-1 is also able to control the accumulation of foreign antigen specific T cells in the lymph nodes through apoptosis which is further mediated by a lower regulation of the gene Bcl-2 PD-L