Learn More
ACROBiosystems ACROBiosystems Rabbit PD-L1 / B7-H1 Protein, His Tag
Catalog No. 501956862
Encompass_Preferred
Recombinant Protein;100UG;Rabbit PD-L1, His Tag (PDL-R52H6) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - His 238 (Accession # G1SUI3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.1 kDa. The protein migrates as 32-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6862 Supplier ACROBiosystems Supplier No. PDLR52H6100UG
Recombinant Protein;100UG;Rabbit PD-L1, His Tag (PDL-R52H6) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - His 238 (Accession # G1SUI3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.1 kDa. The protein migrates as 32-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.