Learn More
ACROBiosystems ACROBiosystems Biotinylated Human PD-L1 / B7-H1 Protein, His,Avitag™
Catalog No. 501956506
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human PD-L1, His,Avitag (PDL-H82E4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Accession # Q9NZQ7-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 16.9 kDa. The protein migrates as 19 kDa and 21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human PD-L1, His,Avitag (Cat. No. PDL-H82E4) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) with a linear range of 0.8-13 ng/mL (QC tested).;Immobilized Biotinylated Human PD-L1, His,Avitag (Cat. No. PDL-H82E4) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-PD-L1 MAb, Human IgG1 with a linear range of 0.2-6 ng/mL (Routinely tested).
Catalog No. 50-195-6506 Supplier ACROBiosystems Supplier No. PDLH82E4200UG
Recombinant Protein;200UG;Biotinylated Human PD-L1, His,Avitag (PDL-H82E4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Accession # Q9NZQ7-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 16.9 kDa. The protein migrates as 19 kDa and 21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human PD-L1, His,Avitag (Cat. No. PDL-H82E4) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) with a linear range of 0.8-13 ng/mL (QC tested).;Immobilized Biotinylated Human PD-L1, His,Avitag (Cat. No. PDL-H82E4) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-PD-L1 MAb, Human IgG1 with a linear range of 0.2-6 ng/mL (Routinely tested).