Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PD-L1 Human Biotin 20ug
SDP

Catalog No. 5000185446
Encompass_Preferred

Recombinant, biotinylated human PD-L1 produced in HEK293 cells and supplied as a lyophilized 20 μg vial for research use. The protein includes C-terminal human Fc and Avi tags, is characterized for purity and molecular mass, and exhibits specific binding activity suitable for PD-1/PD-L1 interaction and immunoassay applications.

  • Expressed in HEK293 mammalian cells.
  • Contains C-terminal human Fc and Avi tags.
  • Site-specific biotinylation via Avi tag.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Predicted molecular mass 54.2 kDa; apparent 70-95 kDa by SDS-PAGE.
  • Demonstrated binding activity with ED50 of 15.30 ng/mL.
  • Store frozen and aliquot after reconstitution for long-term stability.

Catalog No. 50-001-85446 Supplier Medchemexpress LLC Supplier No. HYP7072120UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant, biotinylated human PD-L1 produced in HEK293 cells and supplied as a lyophilized 20 μg vial for research use. The protein includes C-terminal human Fc and Avi tags, is characterized for purity and molecular mass, and exhibits specific binding activity suitable for PD-1/PD-L1 interaction and immunoassay applications.

  • Expressed in HEK293 mammalian cells.
  • Contains C-terminal human Fc and Avi tags.
  • Site-specific biotinylation via Avi tag.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Predicted molecular mass 54.2 kDa; apparent 70-95 kDa by SDS-PAGE.
  • Demonstrated binding activity with ED50 of 15.30 ng/mL.
  • Store frozen and aliquot after reconstitution for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.