Learn More
Medchemexpress LLC PD-L1-IN-1 | 2767424-13-3 | 377.44 | 50 MG
SDP
Catalog No. 5000352066
Encompass_Preferred
PD-L1-IN-1 is a potent PD-L1 inhibitor with an IC50 of 115 nM. It strongly binds to the PD-L1 protein, enhancing the antitumor immune activity of peripheral blood mononuclear cells in co-culture with PD-L1 expressing cancer cells. It increases IFN-γ release and induces apoptosis.
  • Potent PD-L1 inhibitor.
  • Enhances antitumor immune activity.
  • Induces apoptosis.
  • Exhibits low cytotoxicity in healthy cells.
Catalog No. 50-003-52066 Supplier Medchemexpress LLC Supplier No. HY13978150MG
PD-L1-IN-1 is a potent PD-L1 inhibitor with an IC50 of 115 nM. It strongly binds to the PD-L1 protein, enhancing the antitumor immune activity of peripheral blood mononuclear cells in co-culture with PD-L1 expressing cancer cells. It increases IFN-γ release and induces apoptosis.
  • Potent PD-L1 inhibitor.
  • Enhances antitumor immune activity.
  • Induces apoptosis.
  • Exhibits low cytotoxicity in healthy cells.