Learn More
Medchemexpress LLC PD-L1 Mouse Biotin 100ug | 100ug
SDP
Catalog No. 5000187907
Encompass_Preferred
PD-L1 protein serves as a ligand for the inhibitory receptor PDCD1/PD-1 and is critical for maintaining immune tolerance This interaction modulates T cell activation limits effector responses and may stimulate IL10-producing T cell subsets PD-L1 Protein Mouse (Biotinylated HEK293 His) is the recombinant mouse-derived PD-L1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87907 Supplier Medchemexpress LLC Supplier No. HYP77819100UG
PD-L1 protein serves as a ligand for the inhibitory receptor PDCD1/PD-1 and is critical for maintaining immune tolerance This interaction modulates T cell activation limits effector responses and may stimulate IL10-producing T cell subsets PD-L1 Protein Mouse (Biotinylated HEK293 His) is the recombinant mouse-derived PD-L1 protein expressed by HEK293 with C-His labeled tag