Learn More
Medchemexpress LLC PD-L1 Mouse HEK293 100ug
SDP
Catalog No. 5000179708
Encompass_Preferred
PD-L1 protein serves as a ligand for the inhibitory receptor PDCD1/PD-1 and is critical for maintaining immune tolerance This interaction modulates T cell activation limits effector responses and may stimulate IL10-producing T cell subsets PD-L1 Protein Mouse (HEK293 His-Fc) is the recombinant mouse-derived PD-L1 protein expressed by HEK293 with C-hFc C-His labeled tag
Catalog No. 50-001-79708 Supplier Medchemexpress LLC Supplier No. HYP73716100UG
PD-L1 protein serves as a ligand for the inhibitory receptor PDCD1/PD-1 and is critical for maintaining immune tolerance This interaction modulates T cell activation limits effector responses and may stimulate IL10-producing T cell subsets PD-L1 Protein Mouse (HEK293 His-Fc) is the recombinant mouse-derived PD-L1 protein expressed by HEK293 with C-hFc C-His labeled tag