Learn More
Abnova™ PD-L1/PD-L2 Split FISH Probe
Catalog No. 89337273 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
FISH-Ce
Catalog No. For Use With (Application)
89-337-273 FISH-Ce
1 options
Catalog No. 89-337-273 Supplier Abnova™ Supplier No. FS0075
Add to Cart
Add to Cart
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe