Learn More
Sino Biological Recombinant Human PD-L1/B7-H1 Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10084H08HB20
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met1-Thr239) of the extracellular domain of human B7-H1 (NP_054862.1) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6049