Learn More
Sino Biological Recombinant Mouse PD-L1/B7-H1 Protein (ECD, hFc & AVI Tag), Biotinylated
Catalog No. 502115378
Encompass_Preferred
A DNA sequence encoding the mouse CD274 ( NP_068693.1) (Met1-Thr238) was expressed with C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5378 Supplier Sino Biological Supplier No. 50010M41HB100
A DNA sequence encoding the mouse CD274 ( NP_068693.1) (Met1-Thr238) was expressed with C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.