Learn More
Sino Biological Mouse PD-L1 / B7-H1 / CD274 Protein (Fc & AVI Tag), Biotinylated 20ug
Supplier:  Sino Biological 50010M41HB20
Encompass_Preferred
A DNA sequence encoding the mouse CD274 ( NP_068693.1) (Met1-Thr238) was expressed with c-terminal Fc region of human IgG1 tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5364