Learn More
Sino Biological Recombinant Mouse PD-L1/B7-H1 Protein (His Tag), Amine-Labeled Biotinylated 20
Supplier:  Sino Biological 50010M08HB20
Encompass_Preferred
A DNA sequence encoding the Mouse CD274 (NP_068693.1) (Met1-Thr238) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6051