Learn More
ACROBiosystems ACROBiosystems Mouse PD-L2 / B7-DC Protein, Fc Tag
Supplier:  ACROBiosystems PD2M52541MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse PD-L2, Fc Tag (PD2-M5254) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 219 (Accession # NP_067371).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. As a result of glycosylation, the protein migrates as 60-70 kDa under reducing (R) condition, and 120-140 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse PD-L2, Fc Tag (Cat. No. PD2-M5254) at 5 μg/mL (100 μL/well) can bind Mouse PD-1, mouse IgG2a Fc tag (Cat. No. PD1-M52A4) with a linear range of 0.078-0.625 μg/mL (QC tested).
Catalog No. 50-195-7339