Learn More
Medchemexpress LLC PD-L2 Human Biotin 20ug
SDP
Catalog No. 5000183072
Encompass_Preferred
PD-L2 Protein operating independently of PDCD1 plays a vital role in T-cell proliferation and IFNG production Its interaction with PDCD1 inhibits proliferation and cytokine production The interplay between PD-L2 and PDCD1 regulates immune responses influencing T-cell activation and function This molecular interaction adds complexity to mechanisms governing T-cell behavior highlighting PD-L2 s versatile role in immune regulation PD-L2 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived PD-L2 protein expressed by HEK293 with C-Avi C-6 His labeled tag
Catalog No. 50-001-83072 Supplier Medchemexpress LLC Supplier No. HYP7240720UG
PD-L2 Protein operating independently of PDCD1 plays a vital role in T-cell proliferation and IFNG production Its interaction with PDCD1 inhibits proliferation and cytokine production The interplay between PD-L2 and PDCD1 regulates immune responses influencing T-cell activation and function This molecular interaction adds complexity to mechanisms governing T-cell behavior highlighting PD-L2 s versatile role in immune regulation PD-L2 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived PD-L2 protein expressed by HEK293 with C-Avi C-6 His labeled tag