Learn More
Sigma Aldrich Fine Chemicals Biosciences PD Miditrap G 25 Cytiva 28
Catalog No. 502904533
Encompass
PD MidiTrap(TM) G-25 is a prepacked single-use column for buffer exchange and cleanup of biological samples for example proteins and carbohydrates.
Catalog No. 50-290-4533 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28918008
PD MidiTrap(TM) G-25 is a prepacked single-use column for buffer exchange and cleanup of biological samples for example proteins and carbohydrates.