Learn More
Sigma Aldrich Fine Chemicals Biosciences PD MinitrapTM G 10 Cytiva
Catalog No. 502904878
Encompass
PD MiniTrap(TM) G-10 columns are prepacked single-use gravity columns for buffer exchange and cleanup of biological samples larger than Mr 700 for example peptides small proteins and oligosaccharides.
Catalog No. 50-290-4878 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28918010
Add to Cart
Add to Cart
PD MiniTrap(TM) G-10 columns are prepacked single-use gravity columns for buffer exchange and cleanup of biological samples larger than Mr 700 for example peptides small proteins and oligosaccharides.