Learn More
Sigma Aldrich Fine Chemicals Biosciences PD MinitrapTM G 25 Cytiva
Catalog No. 502904760
Encompass
PD MiniTrap(TM) G-25 is a prepacked single-use column for cleanup of large biomolecules (such as proteins 5000 Mr) by removal of buffer salts and low molecular weight compounds.
Catalog No. 50-290-4760 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28918007
Add to Cart
Add to Cart
PD MiniTrap(TM) G-25 is a prepacked single-use column for cleanup of large biomolecules (such as proteins 5000 Mr) by removal of buffer salts and low molecular weight compounds.