Learn More
Crescent Chemical Co Inc PD NITRATE MATRIX MOD 100ML
SDP
Catalog No. 5079653
Encompass_Preferred
Palladium Nitrate, Matrix Modifiers, Concentration: 1% Pd in HNO3, 100mL CAS Size - 100mL Catalog Number - MM-014
Catalog No. 50-796-53 Supplier Crescent Chemical Co Inc Supplier No. IMMM014
Palladium Nitrate, Matrix Modifiers, Concentration: 1% Pd in HNO3, 100mL CAS Size - 100mL Catalog Number - MM-014